Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IL33 (from E. coli)
Human recombinant IL33 (from E. coli)
Catalog # PRSI91-075
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL33 (from E. coli)
Catalog # PRSI91-075
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interleukin-1 Family Member 11|IL-1F11|IL33|C9orf26|IL1F11|NF-HEV|Nuclear Factor From High Endothelial Venules|IL-33|NFHEV|Interleukin-33
  • Protein/peptide name:
    IL33
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    20.29 kD
  • Sequence:
    Ser112-Thr270
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

Interleukin-33 (IL-33) was initially discovered as a nuclear factor NF-HEV abundantly expressed in high endothelial venules. It is a 30-32 kD pro-inflammatory protein with intracellular and extracellular activities and a chromatin-associated cytokine of the IL-1 family with high sequence and structural similarity to IL-1 and IL-18. IL-33 is highly and selectively expressed by high endothelial venule endothelial cells (HEVECs) in human tonsils, Peyers's patches, and lymph nodes. It contains a bipartite nuclear localization signal at the C-terminus, and is targeted to the nucleus when ectopically expressed in human umbilical vein endothelial cells (HUVECs) and HeLa cells. The C-terminal fragment, corresponding to mature IL-33, binds and triggers signaling. IL-33 mediates its biological effects via Toll-interleukin 1 (IL-1) receptor (TIR) domain-containing receptor ST2, activates NF-kappaB and MAP kinases, and drives production of T(H)2-associated cytokines from in vitro polarized T(H)2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. Human IL-33 is 270 amino acids in length.

  • This recombinant protein can be used for biological assays. For research use only.