Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant IL1F10 (fromE. coli)
Human recombinant IL1F10 (fromE. coli)
Catalog # PRSI91-060
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL1F10 (fromE. coli)
Catalog # PRSI91-060
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-1F10|Interleukin-1 HY2|IL-38|FIL1T|Interleukin-1 Theta|Interleukin-1 Family Member 10|FIL1 Theta|IL-1 Theta|IL-1HY2|IL1HY2|IL1F10
  • Protein/peptide name:
    IL1F10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.9 kD
  • Sequence:
    Met1-Trp152
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 6.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Human Interleukin 1 Family Member 10 (IL-1F10) is thought to participate in a network of Interleukin 1 cytokine family members to regulate adapted and innate immune responses. IL-1F10 was expressed in fetal skin, spleen and tonsil, mostly in the basal epithelia of skin and in proliferating B-cells of the tonsil. IL-1F10 binds soluble IL-1 receptor type 1 and may be implicated in regulating adapted and innate immune responses. Two alternatively spliced transcript variants encoding the same protein have been reported.

  • This recombinant protein can be used for biological assays. For research use only.