Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant neuropilin 11 (from HEK293 cells)
Human recombinant neuropilin 11 (from HEK293 cells)
Catalog # PRSI96-562
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant neuropilin 11 (from HEK293 cells)
Catalog # PRSI96-562
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its ability to bind with recombinant human VEGF165 in a functional ELISA.
  • Protein synonyms:
    NRP1|Neuropilin-1|VEGF165R|NRP|CD304
  • Protein/peptide name:
    Neuropilin 11
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    70.7 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Neuropilin-1 (NRP1) is also known as Vascular endothelial cell growth factor 165 receptor (VEGF165R), CD antigen CD304, which belongs to the neuropilin family. The membrane-bound isoform 1 is a receptor involved in the development of the cardiovascular system, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It mediates the chemorepulsant activity of semaphorins. It binds to semaphorin 3A, The PLGF-2 isoform of PGF, The VEGF-165 isoform of VEGF and VEGF-B. Coexpression with KDR results in increased VEGF-165 binding to KDR as well as increased chemotaxis. It may regulate VEGF-induced angiogenesis. The soluble isoform 2 binds VEGF-165 and appears to inhibit its binding to cells.