Human Recombinant CD122 (from HEK293 cells)
Catalog # PRSI96-463
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:CD122|P70-75|IL2RB
- Protein/peptide name:CD122
- Purity:>92% as determined by SDS-PAGE.
- Molecular weight:50.7 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot
Specifications
About this item
Interleukin-2 receptor subunit beta (IL2RB) is also known as CD122, which Belongs to the type I cytokine receptor family. Type 4 subfamily. IL2RB contains 1 fibronectin type-III domain. IL2RB / CD122 is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2.