Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Pecam 11 (from HEK293 cells)
Human recombinant Pecam 11 (from HEK293 cells)
Catalog # PRSI96-093
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Pecam 11 (from HEK293 cells)
Catalog # PRSI96-093
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. When 50000 cells/well are added to rhCD31 coated plates (3. 3 μg/mL, 100 μl/well) in the presence of 20 ng/mL PMA, approximately 55 - 65% will adhere after 30 minutes at 37˚C.
  • Protein synonyms:
    PECAM1|FLJ58394|FLJ34100|CD31
  • Protein/peptide name:
    Pecam 11
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    65.6 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Cluster of Differentiation 31 (CD31) is also known as Platelet endothelial cell adhesion molecule (PECAM-1), is a 130-kDa transmembrane glycoprotein expressed by endothelial cells, platelets, macrophages and Kupffer cells, granulocytes, T / NK cells, lymphocytes, megakaryocytes, osteoclasts, neutrophils, certain tumors, and is the only known member of the CAM family on platelets. CD31 is found on the surface of platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. The encoded protein is a member of the immunoglobulin superfamily and is likely involved in leukocyte migration, angiogenesis, and integrin activation. CD31 plays a key role in removing aged neutrophils from the body. CD31 mediates the homotypic or heterotypic cell adhesion by binding to itself or the leukocyte integrin αvβ3, and thus plays a role in neutrophil recruitment in inflammatory responses, transendothelial migration of leukocytes, as well as in cardiovascular development. In addition, it has been shown that CD31 expression is up-regulated by LPS stimulation, and might function as a feedback negative regulator of LPS inflammatory response in macrophages.