Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:The ED50 was determined by a cell proliferation assay using FDC-P1 cells is = 2.0 ng/mL, corresponding to a specific activity of = 5 x 105 units/mg.
- Protein synonyms:IBP1|IBP3|Mechano growth factor|BP-53|Insulin-like growth factor I|IGF-I
- Protein/peptide name:IGFI
- Purity:>98%
- Formulation:Lyophilized
Specifications
About this item
The IGFs are mitogenic polypeptide growth factors that stimulate the proliferation and survival of various cell types including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar by structure and function to insulin, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but, IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions. Recombinant human IGF-I and IGF-II are globular proteins containing 70 and 67 amino acids, respectively, and 3 intra-molecular disulfide bonds.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.