Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant Psmc6 (from E. coli)
Catalog # PRSIXW-RP3212
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Psmc6 (from E. coli)
Catalog # PRSIXW-RP3212
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    26S protease regulatory subunit 10B|SUG2|P44|26S proteasome AAA-ATPase subunit RPT4|p42|CADP44|HEL-S-73
  • Protein/peptide name:
    Psmc6
  • Purity:
    >95%
  • Molecular weight:
    44.7 kDa (Calculated)
  • Tested applications:
    ELISA, Western Blot, MS

Specifications

About this item

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 α subunits and 2 rings are composed of 7 β subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides.

  • This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.