Order Entry
Czech Republic
ContactUsLinkComponent
Human GBP6 peptide (15 aa near C-terminus)
Human GBP6 peptide (15 aa near C-terminus)
Katalogové číslo PRSI6707P
Dodavatel:  ProSci Inc.
Číslo CAS:  
undefined
Human GBP6 peptide (15 aa near C-terminus)
Katalogové číslo PRSI6707P
Dodavatel:  ProSci Inc.
Číslo CAS:  

Specifikace

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    GBP6 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

Specifikace

O tomto produktu

GBP6 peptide is used for blocking the activity of GBP6 antibody.