Human RUSC2 peptide (15 aa near C-terminus)
PRSI5413P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:RUSC2 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
RUSC2 peptide is used for blocking the activity of RUSC2 antibody.