Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Strumpellin peptide (19 aa near C-terminus)
Catalog # PRSI6497P
Supplier:  ProSci Inc.
CAS Number:  
Human Strumpellin peptide (19 aa near C-terminus)
Catalog # PRSI6497P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa near C-terminus
  • Protein/peptide name:
    Strumpellin Peptide (19 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

Strumpellin peptide is used for blocking the activity of Strumpellin antibody.