Order Entry
Czech Republic
ContactUsLinkComponent
Human FCHSD1 peptide (15 aa near C-terminus)
Human FCHSD1 peptide (15 aa near C-terminus)
  PRSI6469P
 :  
Human FCHSD1 peptide (15 aa near C-terminus)
  PRSI6469P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    FCHSD1 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

NETO1 peptide is used for blocking the activity of NETO1 antibody.