Order Entry
Czech Republic
ContactUsLinkComponent
Chicken apolipoprotein A5 peptide (15 aa near C-terminus)
Chicken apolipoprotein A5 peptide (15 aa near C-terminus)
Catalog # PRSI6271P
Supplier:  ProSci Inc.
CAS Number:  
Chicken apolipoprotein A5 peptide (15 aa near C-terminus)
Catalog # PRSI6271P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Chicken
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    Apolipoprotein A5 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

ApoA5 peptide is used for blocking the activity of ApoA5 antibody.