Order Entry
Czech Republic
ContactUsLinkComponent
Human ZBTB8 peptide (17 aa near C-terminus)
Human ZBTB8 peptide (17 aa near C-terminus)
Catalog # PRSI6129P
Supplier:  ProSci Inc.
CAS Number:  
Human ZBTB8 peptide (17 aa near C-terminus)
Catalog # PRSI6129P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa near C-terminus
  • Protein/peptide name:
    ZBTB8 Peptide (17 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

ZBTB8 peptide is used for blocking the activity of ZBTB8 antibody.