Human KPNA2 peptide (15 aa near N-terminus)
PRSI5983P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near N-terminus
- Protein/peptide name:KPNA2 Peptide (15 aa near N-terminus)
- Tested applications:Blocking
KPNA2 peptide is used for blocking the activity of KPNA2 antibody.