Human ATP11A peptide (19 aa near N-terminus)
PRSI5855P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near N-terminus
- Protein/peptide name:ATP11A Peptide (19 aa near N-terminus)
- Tested applications:Blocking
ATP11A peptide is used for blocking the activity of ATP11A antibody.