Human BCAS4 peptide (14 aa near N-terminus)
PRSI5631P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near N-terminus
- Protein/peptide name:BCAS4 Peptide (14 aa near N-terminus)
- Tested applications:Blocking
BCAS4 pepide is used for blocking the activity of BCAS4 antibody.