Human RSRC1 peptide (19 aa near C-terminus)
Katalogové číslo PRSI5607P
Dodavatel: ProSci Inc.
Číslo CAS:
Specifikace
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near C-terminus
- Protein/peptide name:RSRC1 Peptide (19 aa near C-terminus)
- Tested applications:Blocking
Specifikace
O tomto produktu
RSRC1 pepide is used for blocking the activity of RSRC1 antibody.