Human LXR-A peptide (15 aa near C-terminus)
PRSI5577P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:LXR-A Peptide (15 aa near C-terminus)
- Tested applications:Blocking
LXR-A peptide is used for blocking the activity of LXR-A antibody.