Human PRDM16 peptide (17 aa near C-terminus)
PRSI5555P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near C-terminus
- Protein/peptide name:PRDM16 Peptide (17 aa near C-terminus)
- Tested applications:Blocking
PRDM16 peptide is used for blocking the activity of PRDM16 antibody.