Human PPAPDC1A peptide (13 aa near C-terminus)
Catalog # PRSI5513P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa near C-terminus
- Protein/peptide name:PPAPDC1A Peptide (13 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
PPAPDC1A peptide is used for blocking the activity of PPAPDC1A antibody.