Order Entry
Czech Republic
ContactUsLinkComponent
Human TMEM38A peptide (17 aa near C-terminus)
Human TMEM38A peptide (17 aa near C-terminus)
Katalogové číslo PRSI5521P
Dodavatel:  ProSci Inc.
Číslo CAS:  
undefined
Human TMEM38A peptide (17 aa near C-terminus)
Katalogové číslo PRSI5521P
Dodavatel:  ProSci Inc.
Číslo CAS:  

Specifikace

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa near C-terminus
  • Protein/peptide name:
    TMEM38A Peptide (17 aa near C-terminus)
  • Tested applications:
    Blocking

Specifikace

O tomto produktu

TMEM38A peptide is used for blocking the activity of TMEM38A antibody.