Human GPR3 peptide (13 aa near C-terminus)
Catalog # PRSI5175P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa near C-terminus
- Protein/peptide name:GPR3 Peptide (13 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
GPR3 peptide is used for blocking the activity of GPR3 antibody.