Human Stk39 peptide (18 aa in the center)
PRSI4867P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa in the center
- Protein/peptide name:Stk39 Peptide (18 aa in the center)
- Tested applications:Blocking
Stk39 peptide is used for blocking the activity of Stk39 antibody.