Human CUEDC2 peptide (19 aa near C-terminus)
PRSI4839P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near C-terminus
- Protein/peptide name:CUEDC2 Peptide (19 aa near C-terminus)
- Tested applications:Blocking
CUEDC2 peptide is used for blocking the activity of CUEDC2 antibody.