Human integrin alpha 4# peptide (16 aa in the center)
PRSI4827P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa in the center
- Protein/peptide name:integrin alpha 4# Peptide (16 aa in the center)
- Tested applications:Blocking
Integrin alpha 4 peptide is used for blocking the activity of integrin alpha 4 antibody.