Order Entry
Czech Republic
ContactUsLinkComponent
 
Human ACVR1C peptide (15 aa near N-terminus)
  PRSI4795P
 :  
Human ACVR1C peptide (15 aa near N-terminus)
  PRSI4795P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near N-terminus
  • Protein/peptide name:
    ACVR1C Peptide (15 aa near N-terminus)
  • Tested applications:
    Blocking

 

 

ACVR1C peptide is used for blocking the activity of ACVR1C antibody.