Order Entry
Czech Republic
ContactUsLinkComponent
Mouse TRIM30 peptide (16 aa near C-terminus)
Mouse TRIM30 peptide (16 aa near C-terminus)
Catalog # PRSI4745P
Supplier:  ProSci Inc.
CAS Number:  
Mouse TRIM30 peptide (16 aa near C-terminus)
Catalog # PRSI4745P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Mouse
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa near C-terminus
  • Protein/peptide name:
    TRIM30 Peptide (16 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

TRIM30 peptide is used for blocking the activity of TRIM30 antibody.