Human POLR3F peptide (21 aa near N-terminus)
PRSI4727P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:21 aa near N-terminus
- Protein/peptide name:POLR3F Peptide (21 aa near N-terminus)
- Tested applications:Blocking
POLR3F peptide is used for blocking the activity of POLR3F antibody.