Order Entry
Czech Republic
ContactUsLinkComponent
Human Slc35D1 peptide (20 aa near C-terminus)
Human Slc35D1 peptide (20 aa near C-terminus)
Catalog # PRSI4607P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Slc35D1 peptide (20 aa near C-terminus)
Catalog # PRSI4607P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    20 aa near C-terminus
  • Protein/peptide name:
    Slc35D1 Peptide (20 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

Slc35D1 peptide is used for blocking the activity of Slc35D1 antibody.