Human SAPAP2 peptide (16 aa in the center)
Catalog # PRSI4625P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa in the center
- Protein/peptide name:SAPAP2 Peptide (16 aa in the center)
- Tested applications:Blocking
Specifications
About this item
SAPAP2 peptide is used for blocking the activity of SAPAP2 antibody.