Order Entry
Czech Republic
ContactUsLinkComponent
Human GPAT1 peptide (15 aa near C-terminus)
Human GPAT1 peptide (15 aa near C-terminus)
  PRSI4613P
 :  
undefined
Human GPAT1 peptide (15 aa near C-terminus)
  PRSI4613P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    GPAT1 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

GPAT1 peptide is used for blocking the activity of GPAT1 antibody.