Order Entry
Czech Republic
ContactUsLinkComponent
 
Human cathelicidin peptide (17 aa near C-terminus)
  PRSI4429P
 :  
undefined
Human cathelicidin peptide (17 aa near C-terminus)
  PRSI4429P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 aa near C-terminus
  • Protein/peptide name:
    Cathelicidin Peptide (17 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

Cathelicidin peptide is used for blocking the activity of Cathelicidin antibody.