Order Entry
Czech Republic
ContactUsLinkComponent
 
Human FGF4 peptide (18 aa near C-terminus)
  PRSI4433P
 :  
undefined
Human FGF4 peptide (18 aa near C-terminus)
  PRSI4433P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa near C-terminus
  • Protein/peptide name:
    FGF4 Peptide (18 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

FGF4 peptide is used for blocking the activity of FGF4 antibody.