Order Entry
Czech Republic
ContactUsLinkComponent
Human TEM4 peptide (18 aa in the center)
Human TEM4 peptide (18 aa in the center)
Catalog # PRSI4367P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human TEM4 peptide (18 aa in the center)
Catalog # PRSI4367P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa in the center
  • Protein/peptide name:
    TEM4 Peptide (18 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

TEM4 peptide is used for blocking the activity of TEM4 antibody.