Order Entry
Czech Republic
ContactUsLinkComponent
Human TSC1 peptide (15 aa near C-terminus)
Human TSC1 peptide (15 aa near C-terminus)
Catalog # PRSI3503P
Supplier:  ProSci Inc.
CAS Number:  
Human TSC1 peptide (15 aa near C-terminus)
Catalog # PRSI3503P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near C-terminus
  • Protein/peptide name:
    TSC1 Peptide (15 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

TSC1 peptide is used for blocking the activity of the TSC1 antibody.