Human caspase 4 peptide (16 aa near N-terminus)
PRSI3451P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near N-terminus
- Protein/peptide name:Caspase 4 Peptide (16 aa near N-terminus)
- Tested applications:Blocking
Caspase-4 peptide is used for blocking the activity of caspase-4 antibody.