Human TRIM5 alpha peptide (14 aa in the center)
Catalog # PRSI3251P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:TRIM5 alpha Peptide (14 aa in the center)
- Tested applications:Blocking
Specifications
About this item
TRIM5 alpha peptide is used for blocking the activity of the TRIM5 alpha antibody.