Human ILPIP peptide (13 aa near C-terminus)
PRSI3201P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa near C-terminus
- Protein/peptide name:ILPIP Peptide (13 aa near C-terminus)
- Tested applications:Blocking
ILPIP peptide is used for blocking the activity of the ILPIP antibody.