Mouse TIRAP peptide (15 aa near C-terminus)
PRSI3157P
:
- Pk:50 µG
- Species:Mouse
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:TIRAP Peptide (15 aa near C-terminus)
- Tested applications:Blocking
TIRAP peptide is used for blocking the activity of TIRAP antibody.