Human TIRAP peptide (20 aa in the center)
PRSI3155P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:20 aa in the center
- Protein/peptide name:TIRAP Peptide (20 aa in the center)
- Tested applications:Blocking
TIRAP peptide is used for blocking the activity of TIRAP antibody.