Order Entry
Czech Republic
ContactUsLinkComponent
 
Mouse smac peptide (16 aa near C-terminus)
  PRSI2411P
 :  
undefined
Mouse smac peptide (16 aa near C-terminus)
  PRSI2411P
 :  

 

  • Pk:
    50 µG
  • Species:
    Mouse
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa near C-terminus
  • Protein/peptide name:
    Smac Peptide (16 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

Smac peptide is used for blocking the activity of mSmac/DIABLO antibody.