Mouse RIP3 (14 aa) Peptide
PRSI2283P
:
- Pk:50 µG
- Species:Mouse
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:RIP3 (14 aa) Peptide
- Tested applications:Blocking
RIP3 peptide is used for blocking the activity of RIP3 antibody.