Human GFR alpha 3 peptide (14 aa in the center)
PRSI1137P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:GFR alpha 3 Peptide (14 aa in the center)
- Tested applications:Blocking
GFR alpha 3 peptide is used for blocking the activity of GFR alpha 3 antibody.