Human CXCR4 peptide (15 aa near the extracellualr loop domain)
PRSI1012P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near the extracellualr loop domain
- Protein/peptide name:CXCR4 Peptide (15 aa near the extracellualr loop domain)
- Tested applications:Blocking
CXCR4 peptide is used for blocking the activity of CXCR4 antibody.