Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human AIF peptide

Human AIF peptide

50 µG
Catalog # PRSI2239P
Supplier:   ProSci Inc.
Human AIF peptide
Catalog # PRSI2239P
Supplier:  ProSci Inc.
CAS Number:  
Human AIF peptide

50 µG

CAS Number:  
Price
3 360,00 Kč
Log in for availability
1 * 50 µG
Product Details & Documents

AIF peptide is used for blocking the activity of AIF antibody.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI2239P
  • Protein/peptide name:
    AIF Peptide
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    14 amino acids terminus of human AIF Peptide
  • Tested applications:
    Blocking
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.