Mouse CIDE-A Peptide (15 aa near C-terminus)
Catalog # PRSI2089P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Mouse
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:CIDE-A Peptide (15 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
CIDE-A peptide is used for blocking the activity of mCIDE-A antibody.