Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Drug and metabolite standards, norclobazam, LoGiCal®
Supelco® Drug and metabolite standards, norclobazam, LoGiCal®
Catalog # PROOLGCFOR0910.01
CAS Number:  
Supelco® Drug and metabolite standards, norclobazam, LoGiCal®
Catalog # PROOLGCFOR0910.01
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    22316-55-8
  • Description:
    N-Desmethylclobazam
  • Formula:
    C₁₅H₁₁ClN₂O₂
  • Description:
    N-Desmethylclobazam

Specifications

About this item