Anti-TNFRSF13B Rat Monoclonal Antibody [clone: 1A-10]
PRSI36-145
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 13B
- Antigen symbol:TNFRSF13B
- Clonality:Monoclonal
- Clone:1A-10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:57916
- Antigen synonyms:CD267|Transmembrane activator and CAML interactor|CVID|RYZN|TACI|Tumor necrosis factor receptor superfamily member 13B|CVID2|TNFRSF14B
- Storage buffer:In PBS containing 0.02% sodium azide.
- Storage temperature:Stable for at least 1 year after receipt when stored at -20˚C.
- Concentration:1 mg/ml
- Shipping temperature:-20°C
- Immunogen:Y3 cells transfected with mouse TACI.
- Tested applications:FACS
- Purification:>95% (SDS-PAGE)
- Size:100 μg
- Pk:100 µG
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 1A-10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse