Anti-IL17A Mouse Monoclonal Antibody (FITC) [clone: 17-142F]
Catalog # PRSI36-073
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17A
- Antigen symbol:IL17A
- Clonality:Monoclonal
- Clone:17-142F
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3605
- Antigen synonyms:IL-17|IL17|CTLA8|IL-17A|CTLA-8|Cytotoxic T-lymphocyte-associated serine esterase 8
- Storage buffer:0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage temperature:Stable for at least 1 year after receipt when stored at -20˚C.
- Concentration:1 mg/ml
- Shipping temperature:-20°C
- Immunogen:Recombinant human IL-17A.
- Tested applications:FACS
- Size:50 μg
- Pk:50 µG
Specifications
About this item
IL-17A exhibits multiple biological activities on a variety of cells including the induction of IL-6 and IL-8 production in fibroblsts, the enhancement of surface expression of ICAM-1 in fibroblast, activation of NF-kappaB and costimulation of T cell proliferation. Induces stromal cells to produce proinflammatory and hematopoietic cytokines.
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 17-142F
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human