Anti-IFNG Mouse Monoclonal Antibody [clone: 1.1-5A9-2D05]
Catalog # PRSIXP-5739-M
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Interferon, Gamma
- Antigen symbol:IFNG
- Clonality:Monoclonal
- Clone:1.1-5A9-2D05
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:3458
- Antigen synonyms:AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c x ICR F1 mice using recombinant IFN-gamma as the immunizing antigen.
- Purification:This IgG1k antibody was purified on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 1.1-5A9-2D05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human